4-Epioxytetracycline, 'can be used as secondary standard'
4-Epioxytetracycline, 'can be used as secondary standard'
4-Epioxytetracycline, 'can be used as secondary standard'
4-Epioxytetracycline, 'can be used as secondary standard'
Thermo Scientific Chemicals
4-Epioxytetracycline, 'can be used as secondary standard'
4-Epioxytetracycline, CAS # 14206-58-7, is one of the main oxytetracycline metabolites that show antibacterial activity.
Have Questions?
Catalog NumberQuantityPackaging
ACR257711000100 mgGlass bottle
Catalog number ACR257711000
Price (MYR)
2,072.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,072.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Epioxytetracycline
Name Note97%
CAS14206-58-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
View more
General Description
  • 4-Epioxytetracycline is a metabolite of the oxytetracycline compound that displays antibacterial activity
  • It is the main degradation product derived from the oxytetracycline metabolization and has been detected in some biological samples
  • Its mechanism of action has been described to be related to the mitochondrial protein synthesis inhibition
Applications
  • In laboratory research it has been described that the treatment with 4-epioxytetracycline decreases the bacterial diversity of the intestinal microbiota and facilitates the proliferation of opportunistic pathogens in salmon
  • The use of 4-epioxytetracycline in animal models (wistar rats) resulted in a significant alteration of the gut microbiota and disorders in blood metabolomics
  • 4-Epioxytetracycline can be applied as a secondary reference standard during stability studies of oxytetracycline
  • 4-epioxytetracycline has been used in research studies to evaluate the degradation pathway and products of tetracyclines
RUO – Research Use Only
Literature References: Han, H.; Xiao, H.; Zhang, K.; Lu, Z. Impact of 4-epi-oxytetracycline on the gut microbiota and blood metabolomics of Wistar rats. Scientific reports. 2016, 6, 1-12.Kemper, N. Veterinary antibiotics in the aquatic and terrestrial environment. Ecological indicators. 2008, 8, 1-13.Çelik, A.; Casey, E.; Hasar, H. Degradation of oxytetracycline under autotrophic nitrifying conditions in a membrane aerated biofilm reactor and community fingerprinting. Journal of Hazardous Materials. 2018, 365, 26-33.Harry Björklund.; Johan Bondestam.; Göran Bylund. Residues of oxytetracycline in wild fish and sediments from fish farms.Aquaculture. 1990, 86 (4),359-367.Ole Bent Samuelsen.; Vigdis Torsvik.; Arne Ervik. Long-range changes in oxytetracycline concentration and bacterial resistance towards oxytetracycline in a fish farm sediment after medication.Sci. Total Environ. 1992, 114 25-36.