1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
Thermo Scientific Chemicals
1,10-Dichlorodecane, 98%
CAS: 2162-98-3 | C10H20Cl2 | 211.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR25593025025 gGlass bottle
Catalog number ACR255930250
Price (MYR)
1,452.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,452.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 10-Dichlorodecane
Name Note98%
CAS2162-98-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning