Promethazine hydrochloride
Promethazine hydrochloride
Promethazine hydrochloride
Promethazine hydrochloride
Thermo Scientific Chemicals
Promethazine hydrochloride
CAS: 58-33-3 | C17H20N2S·HCl | 320.88 g/mol
Have Questions?
Catalog NumberQuantity
ACR2545800505 g
Catalog number ACR254580050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPromethazine hydrochloride
CAS58-33-3
Health Hazard 1GHS Signal Word: Warning
Melting Point227.0°C to 230.0°C
EINECS Number200-375-2