Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Thermo Scientific Chemicals
Benzo-18-crown-6, 97%
CAS: 14098-24-9 | C16H24O6 | 312.34 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2439800505 gGlass bottle
Catalog number ACR243980050
Price (MYR)
1,248.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,248.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzo-18-crown-6
CAS14098-24-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Chelation/Complexation reagent
RUO – Research Use Only