Petroleum ether, for analysis, boiling range 60-95°C
Petroleum ether, for analysis, boiling range 60-95°C
Petroleum ether, for analysis, boiling range 60-95°C
Petroleum ether, for analysis, boiling range 60-95°C
Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 60-95°C
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2350700101 LGlass Bottle
Catalog number ACR235070010
Price (MYR)
451.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
451.00
EA
Add to cart
Specifications
Chemical Name or MaterialPetroleum ether
Melting Point-40.0°C
Name NoteBoiling range 60° to 95°C
CAS64742-49-0
ColorColorless
View more
For analysis
RUO – Research Use Only