3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
CAS: 207738-08-7 | C16H22Cl2N2 | 313.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2299900101 gGlass bottle
Catalog number ACR229990010
Price (MYR)
627.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
627.00
BOT
Add to cart
Specifications
Chemical Name or Material3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Name Note98+%
CAS207738-08-7
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning