Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™
Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™
Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™
Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™
Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR2285600252.5 gGlass bottle
Catalog number ACR228560025
Price (MYR)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl-β-D-thiogalactopyranoside
Name Note99+%
CAS367-93-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.