Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Thermo Scientific Chemicals
Saccharin, sodium salt hydrate, 99+%
CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR223372500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSaccharin, sodium salt hydrate
Name Note99+%
CAS82385-42-0
ColorColorless to White
Infrared SpectrumAuthentic