Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Thermo Scientific Chemicals
Acetic anhydride, 99+%, for analysis
CAS: 108-24-7 | C4H6O3 | 102.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR222135000
Price (MYR)
490.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic anhydride
CAS108-24-7
CAS Min %98.0
Health Hazard 1Danger
CAS Max %100.0
View more
Acylation reagent for analysis
RUO – Research Use Only