Benzoyl chloride, 99%, pure, AcroSeal™
Benzoyl chloride, 99%, pure, AcroSeal™
Benzoyl chloride, 99%, pure, AcroSeal™
Benzoyl chloride, 99%, pure, AcroSeal™
Benzoyl chloride, 99%, pure, AcroSeal™
Thermo Scientific Chemicals
Benzoyl chloride, 99%, pure, AcroSeal™
CAS: 98-88-4 | C7H5ClO | 140.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR220271000100 mLAcroSeal™ Glass bottle
Catalog number ACR220271000
Price (MYR)
733.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
733.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzoyl chloride
CAS98-88-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat