Acetyl chloride, 99+%
Acetyl chloride, 99+%
Acetyl chloride, 99+%
Acetyl chloride, 99+%
Thermo Scientific Chemicals
Acetyl chloride, 99+%
Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580 | CAS: 75-36-5 | C2H3ClO | 78.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR219472500
Price (MYR)
545.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
545.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetyl chloride
Name Notep.a.
CAS75-36-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger