Barium carbonate, 99+%, for analysis
Barium carbonate, 99+%, for analysis
Barium carbonate, 99+%, for analysis
Barium carbonate, 99+%, for analysis
Thermo Scientific Chemicals
Barium carbonate, 99+%, for analysis
CAS: 513-77-9 | CBaO3 | 197.34 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR217495000500 gPlastic bottle
Catalog number ACR217495000
Price (MYR)
608.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
608.00
EA
Add to cart
Specifications
Boiling Point1450.0°C
Chemical Name or MaterialBarium carbonate
Melting Point811.0°C
Name Notep.a.
CAS513-77-9