Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR215355000
Price (MYR)
1,247.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,247.00
EA
Add to cart
Specifications
Chemical Name or MaterialSqualane
Acidity0.2mg KOH/g max.
CAS111-01-3
ColorColorless
Density0.8085g/mL