Dichloromethane-d2, for NMR, 99.8 atom % D
Dichloromethane-d2, for NMR, 99.8 atom % D
Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, 99.8 atom % D
CAS: 1665-00-5 | CH2Cl2 | 86.94 g/mol
Have Questions?
Catalog NumberQuantity
ACR21455010010 mL
Catalog number ACR214550100
Price (MYR)
1,130.00
EA
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
1,130.00
EA
Add to cart
Specifications
Chemical Name or MaterialDichloromethane-d2
Density1.3600 g/mL
Refractive Index1.4218
Specific Gravity1.36
Quantity10 mL