Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Thermo Scientific Chemicals
Iodine, 99.5+%, for analysis
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR21249025025 gGlass bottle
Catalog number ACR212490250
Price (MYR)
403.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
BOT
Add to cart
Specifications
Boiling Point185.0°C
Chemical Name or MaterialIodine
Melting Point113.0°C
CAS7553-56-2
Health Hazard 1Danger