Barium oxide, 90%, extra pure, powder
Barium oxide, 90%, extra pure, powder
Barium oxide, 90%, extra pure, powder
Barium oxide, 90%, extra pure, powder
Acros Organics
Barium oxide, 90%, extra pure, powder
CAS: 1304-28-5 | BaO | 153.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR207830250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Boiling Point2000.0°C
Chemical Name or MaterialBarium oxide
Melting Point1920.0°C
CAS1304-28-5
ColorGray to Green-Yellow