Sodium hydroxide, 98.5%, for analysis, pellets
Sodium hydroxide, 98.5%, for analysis, pellets
Sodium hydroxide, 98.5%, for analysis, pellets
Sodium hydroxide, 98.5%, for analysis, pellets
Thermo Scientific Chemicals
Sodium hydroxide, 98.5%, for analysis, pellets
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic Bottle
Catalog number ACR206060025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
ColorWhite
Boiling Point1390.0°C
CAS1310-73-2
Health Hazard 1Danger