L(-)-Epinephrine, 99%
L(-)-Epinephrine, 99%
L(-)-Epinephrine, 99%
L(-)-Epinephrine, 99%
Thermo Scientific Chemicals
L(-)-Epinephrine, 99%
CAS: 51-43-4 | C9H13NO3 | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR204400050
Price (MYR)
928.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
928.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(-)-Epinephrine
Name Note99%
CAS51-43-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation. Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure.