Ammonium fluoride, 98+%, extra pure
Ammonium fluoride, 98+%, extra pure
Ammonium fluoride, 98+%, extra pure
Ammonium fluoride, 98+%, extra pure
Thermo Scientific Chemicals
Ammonium fluoride, 98+%, extra pure
CAS: 12125-01-8 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR201845000
Price (MYR)
1,098.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,098.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium fluoride
Name Notep.a.
CAS12125-01-8
ColorColorless or White
Health Hazard 1GHS Signal Word: Danger