Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR201672500250 gPlastic bottle
Catalog number ACR201672500
Price (MYR)
949.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
949.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNickel
Name Notep.a.
CAS7440-02-0
Melting Point1455.0°C
Boiling Point2732.0°C to 2837.0°C
View more
For ultra trace analysis
RUO – Research Use Only