DL-1,2-Hexanediol, 98%
DL-1,2-Hexanediol, 98%
DL-1,2-Hexanediol, 98%
DL-1,2-Hexanediol, 98%
Thermo Scientific Chemicals
DL-1,2-Hexanediol, 98%
CAS: 6920-22-5 | C6H14O2 | 118.18 g/mol
Have Questions?
Catalog NumberQuantity
ACR200132500250 g
Catalog number ACR200132500
Price (MYR)
870.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
870.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-1, 2-Hexanediol
CAS6920-22-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.