Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Thermo Scientific Chemicals
Hydrazine hydrate, 100% (Hydrazine, 64%)
CAS: 10217-52-4 | H4N2 | 32.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR196715000
Price (MYR)
779.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
779.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrazine hydrate
Name NoteHydrazine Hydrate 100% (Hydrazine, 64%)
CAS10217-52-4
ColorColorless
Health Hazard 1Danger
View more
Reduction reagent
RUO – Research Use Only