Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
CAS: 10031-53-5 | EuN3O9·6H2O | 446.07 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR196680050
5 g
Glass bottle
Catalog number ACR196680050
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium(III) nitrate hexahydrate
Name Note99.9%
CAS10031-53-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: May intensify fire; oxidizer.