Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Thermo Scientific Chemicals
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Catalog NumberQuantity
ACR196205000500 mL
Catalog number ACR196205000
Price (MYR)
550.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
550.00
BOT
Add to cart
Chemical Identifiers
CAS7440-09-7, 7732-18-5
Specifications
Chemical Name or MaterialPotassium standard solution
Name Note1 mg/ml K in 2% HNO3
PackagingPlastic bottle
CAS7440-09-7
Health Hazard 1GHS Signal Word: Warning
View more
for atomic adsorption spectroscopy
RUO – Research Use Only