Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Thermo Scientific Chemicals
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR196125000
Price (MYR)
1,043.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,043.00
EA
Add to cart
Chemical Identifiers
CAS7783-34-8, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialMercury standard solution
Name Note1 mg/ml Hg in 10% HNO3
PackagingPlastic bottle
CAS7783-34-8
Health Hazard 1Warning
View more
For atomic adsorption spectroscopy
RUO – Research Use Only