Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Thermo Scientific Chemicals
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Catalog NumberQuantity
ACR196105000500 mL
Catalog number ACR196105000
Price (MYR)
686.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
686.00
BOT
Add to cart
Chemical Identifiers
CAS7439-95-4, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialMagnesium standard solution
Name Note1 mg/ml Mg in 0.5N HNO3
PackagingPlastic bottle
CAS7439-95-4
Health Hazard 1GHS Signal Word: Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only