Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Thermo Scientific Chemicals
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
CAS: 7440-38-2 | As | 74.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR195825000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-38-2, 1310-58-3, 7732-18-5
Specifications
Chemical Name or MaterialArsenic standard solution
Name Note1 mg/ml As in 2% KOH
PackagingPlastic bottle
CAS7440-38-2
Health Hazard 1GHS Signal Word: Danger
View more
for atomic adsorption spectroscopy
RUO – Research Use Only