Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol
Have Questions?
Catalog number ACR194620500
Price (MYR)
1,294.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
1,294.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPraseodymium(III, IV) oxide
Name Note99.9%
CAS12037-29-5
ColorBlack or Brown to Green
Health Hazard 1GHS Signal Word: Warning
View more
for ultra trace analysis
RUO – Research Use Only