Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Platinum, 99.999%, (trace metal basis), powder
Thermo Scientific Chemicals
Platinum, 99.999%, (trace metal basis), powder
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1937100101 gGlass bottle
Catalog number ACR193710010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum
ColorGray
CAS7440-06-4
Melting Point1772.0°C
Boiling Point3827.0°C
View more
For ultra trace analysis
RUO – Research Use Only