Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
Thermo Scientific Chemicals
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1934000101 gGlass bottle
Catalog number ACR193400010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLutetium(III) oxide
Name Note99.99%, -325 mesh
CAS12032-20-1
ColorWhite
Health Hazard 1HYGROSCOPIC
View more
for ultra trace analysis
RUO – Research Use Only