Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Poly(propylene glycol), average M.W. 425
Acros Organics
Poly(propylene glycol), average M.W. 425
CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR192150050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(propylene glycol)
Name NoteAverage M.W. 425
CAS25322-69-4
Boiling Point200.0°C
Melting Point-25.0°C