Sudan II, pure
Sudan II, pure
Sudan II, pure
Thermo Scientific Chemicals
Sudan II, pure
CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol
Have Questions?
Catalog NumberQuantity
ACR19015025025 g
Catalog number ACR190150250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSudan II
Melting Point156.0°C to 158.0°C
CAS3118-97-6
Health Hazard 1CANCER SUSPECTED AGENT
Health Hazard 2GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.