Mercurochrome, 24-27% Mercury
Mercurochrome, 24-27% Mercury
Mercurochrome, 24-27% Mercury
Mercurochrome, 24-27% Mercury
Thermo Scientific Chemicals
Mercurochrome, 24-27% Mercury
CAS: 129-16-8 | C20H8Br2HgNa2O6 | 750.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR190111000
Price (MYR)
1,189.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,189.00
EA
Add to cart
Specifications
Chemical Name or MaterialMercurochrome
CAS129-16-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.