Check Order Status
Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™
Element Search
Structure Search
Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™
Have Questions?
Catalog Number
Quantity
Packaging
ACR183301000
100 g
Glass bottle
Catalog number
ACR183301000
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Poly(vinyl alcohol)
Color
White
CAS
9002-89-5
Physical Form
Powder or Granules
Ash
1% max.
View more
Indicator