Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Thermo Scientific Chemicals
Poly(vinyl acetate), approx. M.W. 100,000
CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR183265000500 gGlass bottle
Catalog number ACR183265000
Price (MYR)
1,127.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,127.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl acetate)
Name NoteAverage M.W. 101600
CAS9003-20-7
Physical FormBeads
Molecular Formula(C4H6O2)n
View more
Suitable for battery materials development.
RUO – Research Use Only