Ethyl chloroformate, 97%
Ethyl chloroformate, 97%
Ethyl chloroformate, 97%
Ethyl chloroformate, 97%
Thermo Scientific Chemicals
Ethyl chloroformate, 97%
CAS: 541-41-3 | C3H5ClO2 | 108.52 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR181081000100 mLGlass bottle
Catalog number ACR181081000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl chloroformate
CAS541-41-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Fatal if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Coupling reagent
RUO – Research Use Only