2-Naphthol, 99+%
2-Naphthol, 99+%
2-Naphthol, 99+%
Thermo Scientific Chemicals
2-Naphthol, 99+%
CAS: 135-19-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number ACR180825000
Price (MYR)
955.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
955.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Naphthol
Name Note99+%
CAS135-19-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning