Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Thermo Scientific Chemicals
Boric acid, 99+%, extra pure
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1805700101 kgPlastic bottle
Catalog number ACR180570010
Price (MYR)
618.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
618.00
EA
Add to cart
Specifications
Chemical Name or MaterialBoric acid
ColorWhite
CAS10043-35-3
Melting Point169.0°C
Health Hazard 1Danger
View more
Lewis acid
RUO – Research Use Only