Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Thermo Scientific Chemicals
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Catalog NumberQuantity
ACR180301000100 g
Catalog number ACR180301000
Price (MYR)
419.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
419.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl alcohol)
CAS9002-89-5
Melting Point230.0°C to 240.0°C
pH4.5 to 6.5 (4% aq. soln.)
Infrared SpectrumAuthentic