Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Thermo Scientific Chemicals
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
CAS: 32001-55-1 | C5H6FN | 99.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR180071000100 gPlastic Bottle in Tin Can
Catalog number ACR180071000
Price (MYR)
1,145.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle in Tin Can
Request bulk or custom format
Price (MYR)
1,145.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrogen fluoride-pyridine
Name Note65-70%
CAS32001-55-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
View more
C-X bond formation reagent
RUO – Research Use Only