Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Thermo Scientific Chemicals
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR17878025025 gGlass bottle
Catalog number ACR178780250
Price (MYR)
1,353.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,353.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(methyl methacrylate)
ColorWhite
Name NoteM.W. 93300
CAS9011-14-7
Physical FormBeads