Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Thermo Scientific Chemicals
Poly(methyl methacrylate), beads, average M.W. 35,000
CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
View all
Glass bottle
Catalog number ACR178765000
Price (MYR)
1,445.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,445.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(methyl methacrylate)
ColorWhite
Name Notenominal M.W. 38000
CAS9011-14-7
Physical FormBeads