Acetone, 99+%, extra pure
Acetone, 99+%, extra pure
Acetone, 99+%, extra pure
Acetone, 99+%, extra pure
Thermo Scientific Chemicals
Acetone, 99+%, extra pure
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1771700505 LPlastic Drum
Catalog number ACR177170050
Price (MYR)
767.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
767.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
CAS67-64-1
CAS Min %99.0
Health Hazard 1Danger