Toluene, 99+%, extra pure
Toluene, 99+%, extra pure
Toluene, 99+%, extra pure
Toluene, 99+%, extra pure
Thermo Scientific Chemicals
Toluene, 99+%, extra pure
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1771600101 LGlass Bottle
Catalog number ACR177160010
Price (MYR)
382.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
382.00
EA
Add to cart
Specifications
Chemical Name or MaterialToluene
Melting Point-95.0°C
CAS108-88-3
Health Hazard 1Signal Word: DANGER!
Health Hazard 2GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag