Acetone, 99.5+%, for analysis
Acetone, 99.5+%, for analysis
Acetone, 99.5+%, for analysis
Acetone, 99.5+%, for analysis
Thermo Scientific Chemicals
Acetone, 99.5+%, for analysis
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Plastic Bottle
Glass Bottle
Catalog number ACR176800026
Price (MYR)
656.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
656.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
Name NoteFor analysis
CAS67-64-1
CAS Min %99.5
View more
For analysis
RUO – Research Use Only