Chloroacetone, 96%, stabilized
Chloroacetone, 96%, stabilized
Chloroacetone, 96%, stabilized
Chloroacetone, 96%, stabilized
Thermo Scientific Chemicals
Chloroacetone, 96%, stabilized
CAS: 78-95-5 | C3H5ClO | 92.52 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR17347025025 mLGlass bottle
Catalog number ACR173470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroacetone
Name NoteStabilized
CAS78-95-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Causes ski