3-Nitrophenol, 99%
3-Nitrophenol, 99%
3-Nitrophenol, 99%
Thermo Scientific Chemicals
3-Nitrophenol, 99%
CAS: 554-84-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR17230010010 gGlass bottle
Catalog number ACR172300100
Price (MYR)
662.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
662.00
EA
Add to cart
Specifications
Chemical Name or Material3-Nitrophenol
CAS554-84-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear prote