DL-Tryptophan, 98%
DL-Tryptophan, 98%
DL-Tryptophan, 98%
Thermo Scientific Chemicals
DL-Tryptophan, 98%
CAS: 54-12-6 | C11H12N2O2 | 204.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR172111000100 gPlastic Bottle
Catalog number ACR172111000
Price (MYR)
1,085.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,085.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-Tryptophan
Name Note99%
CAS54-12-6
ColorBeige to White or Yellow
Infrared SpectrumAuthentic