Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, CAS # 69-93-2, is a common oxopurine metabolite, and a normal component of urine.
Have Questions?
Catalog NumberQuantityPackaging
ACR17129025025 gPlastic bottle
Catalog number ACR171290250
Price (MYR)
467.00
EA
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
467.00
EA
Add to cart
Specifications
Chemical Name or MaterialUric acid
Name Note99+%
CAS69-93-2
ColorWhite
RTECS NumberYU7050080
View more
General description
• Uric acid is a purine-based metabolite found in humans, mice and E. coli
• Uric acid can accumulate in excess leading to diseases such as hyperuricemia
• Effects of excess uric acid accumulation include hypertension, arthritis, and renal issues
Applications
• Useful as a biochemical standard
• Suitable for use in the study of hyperuricemia and gout
• Uric acid may also play other roles in cell metabolism and physiology
• Intended for research use only
RUO – Research Use Only
General References:
  1. Chen, M.; Lu, X.; Lu, C.; Shen, N.; Jiang, Y.; Chen, M.; Wu, H. Soluble uric acid increases PDZK1 and ABCG2 expression in human intestinal cell lines via the TLR4-NLRP3 inflammasome and PI3K/Akt signaling pathway. Arthritis Res Ther. 2018 20(1), 20.
  2. Al Abdulmonem, W.; Aljohani, A. S. M.; Alhumaydhi, F. A.; Mousa, A. H. M.; Rasheed, Z. Protective Potential of Uric Acid, Folic Acid, Glutathione and Ascorbic Acid Against the Formation of Toxic Met-Myoglobin. Protein Pept Lett. 2021 28(3), 282-289.