Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog number ACR169020250
Price (MYR)
2,186.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
2,186.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Density1.1900g/mL
Deuterium (D)99.95% min.
Flash Point88°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only